missing translation for 'onlineSavingsMsg'
Learn More

2-(Trifluoromethyl)benzylamine, 98%, Thermo Scientific™

Product Code. 11470288
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470288 2 g 2g
11480288 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470288 Supplier Thermo Scientific Alfa Aesar Supplier No. B23822.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3048-01-9
Molecular Formula C8H8F3N
Molecular Weight (g/mol) 175.15
MDL Number MFCD00010297
InChI Key ZSKQIFWUTUZAGF-UHFFFAOYSA-N

Specifications

Density 1.249
Boiling Point 108°C to 110°C (64 mmHg)
Flash Point 69°C (156°F)
Refractive Index 1.471
Quantity 2 g
UN Number UN2735
Beilstein 1103424
Sensitivity Air Sensitive
Synonym 2-trifluoromethyl benzylamine, 2-trifluoromethylbenzylamine, 2-trifluoromethyl phenyl methanamine, o-trifluoromethylbenzyl amine, o-trifluoromethylbenzylamine, 2-trifluoromethyl-benzylamine, 1-2-trifluoromethyl phenyl methanamine, benzenemethanamine, 2-trifluoromethyl, o-trifluoromethyl benzylamine
SMILES C1=CC=C(C(=C1)CN)C(F)(F)F
IUPAC Name [2-(trifluoromethyl)phenyl]methanamine
PubChem CID 76447
Formula Weight 175.15
Percent Purity 98%
Chemical Name or Material 2-(Trifluoromethyl)benzylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.