missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human 20S Immunoproteasome Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 413.00 €
Specifications
| Antigen | 20S Immunoproteasome |
|---|---|
| Clone | 894510 |
| Dilution | Immunocytochemistry 5-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749548
![]() |
R&D Systems
MAB8194-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747818
![]() |
R&D Systems
MAB8194-100 |
100 μg |
413.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
20S Immunoproteasome Monoclonal specifically detects 20S Immunoproteasome in Human samples. It is validated for Immunocytochemistry.Specifications
| 20S Immunoproteasome | |
| Immunocytochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 894510 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Human 20S Immunoproteasome purified from peripheral blood mononuclear cells | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human 20S Immunoproteasome in direct ELISA. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title