missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NDUFB5 Polyclonal antibody specifically detects NDUFB5 in Human samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | NDUFB5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | 5 (16kD, SGDH), CISGDH, CI-SGDH, Complex I-SGDH, DKFZp686N02262, FLJ30597, MGC111204, MGC12314, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial, SGDH |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 100-189 of human NDUFB5 (NP_002483.1).,, Sequence:, EIPEGYVPEHWEYYKHPISRWIARNFYDSPEKIYERTMAVLQIEAEKAELRVKELEVRKLMHVRGDGPWYYYETIDKELIDHSPKATPDN |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?