missing translation for 'onlineSavingsMsg'
Learn More

21-Chloro-9-fluoro-11beta,17-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione (Clobetasol), Mikromol™

Product Code. 18080670
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18080670 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18080670 Supplier Mikromol Supplier No. MM0200.01
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

21-Chloro-9-fluoro-11beta,17-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione (Clobetasol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Clobetasol
CAS 25122-41-2
Analyte or Component Names Clobetasol
Shipping Condition Ice pack (-20°C)
Molecular Formula C22 H28 Cl F O4
Certifications/Compliance ISO 17025
Synonym Clobetasol, 21-Chloro-9-fluoro-11beta,17-dihydroxy-16beta-methylpregna-1,4-diene-3,20-dione, Clobetasol Propionate Imp. G (EP), Betamethasone Imp. B (EP), Ph Eur Clobetasol Propionate Impurity G, CCI 4650, Clobetasol, CCI18747, CCI6990, CCI1822, USP Clobetasol propionate Related Compound A, Betamethasone Impurity B, Clobetasol Propionate Impurity G
InChI Formula InChI=1S/C22H28ClFO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,24)17(26)10-20(16,3)22(12,28)18(27)11-23/h6-7,9,12,15-17,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CCl
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-chloroacetyl)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 410.91
Quantity 100 mg
Formula Weight 410.166 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.