Learn More
Human VCAM-1/CD106 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11671-SP
 View more versions of this product
Product Code. 30306744
  • 232.00 € / 25µg
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
VCAM-1/CD106 Monoclonal antibody specifically detects VCAM-1/CD106 in Human samples. It is validated for Immunohistochemistry,Multiplex Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VCAM-1/CD106
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
CD106, CD106 antigen, DKFZp779G2333, INCAM-100, L1CAM, MGC99561, vascular cell adhesion molecule 1, vascular cell adhesion protein 1, V-CAM 1, VCAM1, VCAM-1
Chinese Hamster Ovary cell line, CHO-derived human VCAM-1/CD106, Phe25-Glu698, Accession # P19320
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Immunohistochemistry, Multiplex Immunofluorescence
1027406
Immunohistochemistry 3-25 μg/mL, Multiplex Immunofluorescence 15 μg/mL
P19320
Mouse
Protein A or G purified from cell culture supernatant
RUO
7412
Human
Antibody
IgG1