missing translation for 'onlineSavingsMsg'
Learn More

(2R)-2-Amino-2-phenylacetic Acid (D-Phenylglycine), Mikromol™

Product Code. 30462820
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462820 4 x 25 mg 100mg
30051822 25 mg 25mg
17939484 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30462820 Supplier Mikromol Supplier No. MM0253.1200254AMP
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2R)-2-Amino-2-phenylacetic Acid (D-Phenylglycine), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-2-Amino-2-phenylacetic Acid (D-Phenylglycine)
CAS 875-74-1
Analyte or Component Names (2R)-2-Amino-2-phenylacetic Acid (D-Phenylglycine)
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O2
Certifications/Compliance ISO 17025
Synonym Ampicillin Sodium Imp. L (EP), Ampicillin Imp. L (EP), Cefaclor Imp. A (EP), Cefalexin Monohydrate Imp. A (EP), Cefalexin Imp. A (EP), (2R)-2-Amino-2-phenylacetic Acid, D-Phenylglycine, Ampicillin Sodium Impurity L, Cefaclor Impurity A, Cefalexin Monohydrate Impurity A, Ampicillin Impurity L, Cefalexin Impurity A
InChI Formula InChI=1S/C8H9NO2/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H,9H2,(H,10,11)/t7-/m1/s1
Solution Type Neat
SMILES N[C@@H](C(=O)O)c1ccccc1
IUPAC Name (2R)-2-amino-2-phenylacetic acid
Molecular Weight (g/mol) 151.16
Quantity 4 x 25 mg
Formula Weight 151.0633 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.