missing translation for 'onlineSavingsMsg'
Learn More

(2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride, Mikromol

Product Code. 30461464
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30461464 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461464 Supplier Mikromol Supplier No. MM0455.320025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride
Analyte or Component Names (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C15 H14 Cl N O2 S . H Cl
Certifications/Compliance ISO 9001
Synonym (2R)-(2-Chlorophenyl)[6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl]acetic Acid Hydrochloride, (R)-Clopidogrel Carboxylic Acid Hydrochloride
InChI Formula InChI=1S/C15H14ClNO2S.ClH/c16-12-4-2-1-3-11(12)14(15(18)19)17-7-5-13-10(9-17)6-8-20-13;/h1-4,6,8,14H,5,7,9H2,(H,18,19);1H/t14-;/m1./s1
Solution Type Neat
SMILES Cl.OC(=O)[C@H](N1CCc2sccc2C1)c3ccccc3Cl
IUPAC Name (2R)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetic acid;hydrochloride
Molecular Weight (g/mol) 344.256
Quantity 25 mg
Formula Weight 343.02 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.