missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride, Mikromol™
Description
(2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[(2-(3,4-dimethoxyphenyl)-ethyl)(methyl)amino]-2-ethylpentanenitrile Hydrochloride |
| CAS | 190850-49-8 |
| Analyte or Component Names | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[(2-(3,4-dimethoxyphenyl)-ethyl)(methyl)amino]-2-ethylpentanenitrile Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H36 N2 O4 . HCl |
| Certifications/Compliance | ISO 17025 |
| Synonym | Verapamil Hydrochloride Imp. H (EP), Verapamil Imp. H (EP), (2RS)-2-(3,4-Dimethoxyphenyl)-5-[[2-(3,4-dimethoxyphenyl)-ethyl](methyl)amino]-2-ethylpentanenitrile Hydrochloride, Verapamil Hydrochloride Imp. H (EP) as Hydrochloride, Verapamil Hydrochloride Impurity H as Hydrochloride, Verapamil Impurity H as Hydrochloride |
| InChI Formula | InChI=1S/C26H36N2O4.ClH/c1-7-26(19-27,21-10-12-23(30-4)25(18-21)32-6)14-8-15-28(2)16-13-20-9-11-22(29-3)24(17-20)31-5;/h9-12,17-18H,7-8,13-16H2,1-6H3;1H |
| SMILES | Cl.CCC(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2 |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-ethylpentanenitrile;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?