missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride, Mikromol™

Product Code. 17913276
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17913276 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17913276 Supplier Mikromol Supplier No. MM0022.100025
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride
CAS 67775-97-7
Analyte or Component Names (2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C17 H26 N2 O2 . Cl H
Certifications/Compliance ISO 17025
Synonym Verapamil Hydrochloride Imp. F (EP), Verapamil Imp. F (EP), Verapamil USP Related Compound A, Verapamil USP RC A, (2RS)-2-(3,4-Dimethoxyphenyl)-5-(methylamino)-2-(1-methylethyl)pentanenitrile Hydrochloride, Verapamil Hydrochloride Imp. F (EP) as Hydrochloride, Verapamil Related Compound A as Hydrochloride, Verapamil Hydrochloride Impurity F as Hydrochloride, Verapamil Impurity F as Hydrochloride
InChI Formula InChI=1S/C17H26N2O2.ClH/c1-13(2)17(12-18,9-6-10-19-3)14-7-8-15(20-4)16(11-14)21-5;/h7-8,11,13,19H,6,9-10H2,1-5H3;1H
SMILES Cl.N#CC(C1=CC=C(OC)C(OC)=C1)(CCCNC)C(C)C
IUPAC Name 2-(3,4-dimethoxyphenyl)-5-(methylamino)-2-propan-2-ylpentanenitrile;hydrochloride
Molecular Weight (g/mol) 326.86
Quantity 25 mg
Formula Weight 326.1761 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.