missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic Acid (rac-Carbidopa), Mikromol™

Product Code. 30052259
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052259 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052259 Supplier Mikromol Supplier No. MM1501.120025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic Acid (rac-Carbidopa)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D,L Carbidopa
CAS 302-53-4
Analyte or Component Names D,L Carbidopa
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2 O4
Certifications/Compliance ISO 9001
Synonym Benzenepropanoic acid, α-hydrazinyl-3,4-dihydroxy-α-methyl-, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl- (9CI), Hydrocinnamic acid, α-hydrazino-3,4-dihydroxy-α-methyl- (7CI,8CI), α-Hydrazinyl-3,4-dihydroxy-α-methylbenzenepropanoic acid, DL-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid, DL-Carbidopa, DL-α-Hydrazino-3,4-dihydroxy-α-methylhydrocinnamic acid, DL-α-Hydrazino-α-methyl-3,4-dihydroxyphenylpropionic acid, DL-α-Hydrazino-α-methyl-β-(3,4-dihydroxyphenyl)propionic acid, DL-α-Hydrazino-α-methyldopa, DL-α-MDH, NSC 92521, dl-MK 485, dl-α-Methyl-α-hydrazino-3,4-dihydroxyphenylpropionic acid, α-Hydrazino-α-methyl-3,4-dihydroxyphenylpropionic acid, α-Hydrazino-α-methyldopa, α-MDH, α-Methylhydrazinodopa, β-(3,4-Dihydroxyphenyl)-α-hydrazino-α-methylpropionic acid, β-(3,4-Dihydroxyphenyl)-α-methyl-α-hydrazinopropionic acid
InChI Formula InChI=1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)
SMILES CC(Cc1ccc(O)c(O)c1)(NN)C(=O)O
IUPAC Name 3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
Molecular Weight (g/mol) 226.23
Quantity 25 mg
Formula Weight 226.0954 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.