missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid, Mikromol™

Product Code. 30050468
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050468 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050468 Supplier Mikromol Supplier No. MM0894.150025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid
CAS 151262-57-6
Analyte or Component Names 3-[(4-Fluorophenyl)sulfonyl]-2-hydroxy-2-methylpropanoic Acid
Shipping Condition Room Temperature
Molecular Formula C10H11FO5S
Certifications/Compliance ISO 9001
Synonym Bicalutamide EP Impurity M
SMILES CC(O)(CS(=O)(=O)c1ccc(F)cc1)C(=O)O
Molecular Weight (g/mol) 262.25
Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.