missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-3-Hydroxy-2-phenylpropanoic Acid (Tropic Acid), Mikromol™

Product Code. 17923576
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17923576 25 mg 25mg
17931996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17923576 Supplier Mikromol Supplier No. MM0601.030025
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-3-Hydroxy-2-phenylpropanoic Acid (Tropic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rac Tropic acid
CAS 552-63-6
Analyte or Component Names rac Tropic acid
Shipping Condition Room Temperature
Molecular Formula C9 H10 O3
Certifications/Compliance ISO 17025
Synonym Atropine Imp. C (EP), Hyoscine Butylbromide Imp. B (EP), Hyoscine Hydrobromide Imp. D (EP), Hyoscyamine Sulfate Imp. A (EP), Ipratropium Bromide Imp. C (EP), Tropicamide Imp. C (EP), Ipratropium Bromide USP Related Compound C, Ipratropium Bromide USP RC C, (2RS)-3-Hydroxy-2-phenylpropanoic Acid, dl-Tropic Acid, Tropic Acid, Hyoscyamine Sulphate Imp. A (EP), (+/-)-alpha-(Hydroxymethyl)benzeneacetic acid, (+/-)-2-Phenyl-3-hydroxypropionic acid, (+/-)-3-Hydroxy-2-phenylpropionic acid, (+/-)-Tropic acid, 2-Phenyl-3-hydroxypropanoic acid, 2-Phenyl-3-hydroxypropionic acid, 2-Phenylhydracrylic acid, 3-Hydroxy-2-phenylpropionic acid, DL-Tropic acid, NSC 20990, alpha-(Hydroxymethyl)benzeneacetic acid, Ipratropium Related Compound C, Tropicamide Related Compound C, Atropine Impurity C, Hyoscine Butylbromide Impurity B, Hyoscine Hydrobromide Impurity D, Hyoscyamine Sulfate Impurity A, Hyoscyamine Sulfate Impurity A, Ipratropium Bromide Impurity C, Tropicamide Impurity C, Hyoscyamine Impurity A, Ipratropium Impurity C
InChI Formula InChI=1S/C9H10O3/c10-6-8(9(11)12)7-4-2-1-3-5-7/h1-5,8,10H,6H2,(H,11,12)
SMILES OCC(C(=O)O)c1ccccc1
IUPAC Name 3-hydroxy-2-phenylpropanoic acid
Molecular Weight (g/mol) 166.17
Quantity 25 mg
Formula Weight 166.063 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.