missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride (Norpromethazine Hydrochloride), Mikromol™

Product Code. 17962966
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962966 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962966 Supplier Mikromol Supplier No. MM0412.03
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride (Norpromethazine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride (Norpromethazine Hydrochloride)
CAS 60113-77-1
Analyte or Component Names (2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride (Norpromethazine Hydrochloride)
Shipping Condition Room Temperature
Molecular Formula C16 H18 N2 S . Cl H
Certifications/Compliance ISO 17025
Synonym Promethazine Hydrochloride Imp. C (EP), Promethazine Imp. C (EP), (2RS)-N-Methyl-1-(10H-phenothiazin-10-yl)propan-2-amine Hydrochloride, Promethazine Hydrochloride Imp. C (EP) as Hydrochloride, Norpromethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine, N,alpha-dimethyl-, hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine, N,alpha-dimethyl-, monohydrochloride (9CI), Promethazine Hydrochloride Impurity C as Hydrochloride, Promethazine Impurity C as Hydrochloride
InChI Formula InChI=1S/C16H18N2S.ClH/c1-12(17-2)11-18-13-7-3-5-9-15(13)19-16-10-6-4-8-14(16)18;/h3-10,12,17H,11H2,1-2H3;1H
SMILES Cl.CNC(C)CN1c2ccccc2Sc3ccccc13
IUPAC Name N-methyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
Molecular Weight (g/mol) 306.85
Quantity 50 mg
Formula Weight 306.0957 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.