missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-[[(3S)-1-(Carboxymethyl)-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl]amino]-4-phenylbutanoic Acid (Benazeprilat), Mikromol™

Product Code. 17993006
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17993006 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17993006 Supplier Mikromol Supplier No. MM0274.010025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-[[(3S)-1-(Carboxymethyl)-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl]amino]-4-phenylbutanoic Acid (Benazeprilat)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benazeprilat
CAS 86541-78-8
Analyte or Component Names Benazeprilat
Shipping Condition Room Temperature
Molecular Formula C22 H24 N2 O5
Certifications/Compliance ISO 17025
Synonym Benazepril Hydrochloride Imp. C (EP), Benazepril Imp. C (EP), Benazepril USP Related Compound C, Benazepril USP RC C, (2S)-2-[[(3S)-1-(Carboxymethyl)-2-oxo-2,3,4,5-tetrahydro-1H-1-benzazepin-3-yl]amino]-4-phenylbutanoic Acid (Benazeprilat), 1H-1-Benzazepine-1-acetic acid, 3-[[(1S)-1-carboxy-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-, (3S)-, 1H-1-Benzazepine-1-acetic acid, 3-[(1-carboxy-3-phenylpropyl)amino]-2,3,4,5-tetrahydro-2-oxo-, [S-(R*,R*)]-, Benazeprilat, Benazeprilate, CGS 14831, Benazepril Related Compound C (USP), Benazepril Related Compound C, Benazepril Hydrochloride Impurity C, Benazepril Impurity C
InChI Formula InChI=1S/C22H24N2O5/c25-20(26)14-24-19-9-5-4-8-16(19)11-13-17(21(24)27)23-18(22(28)29)12-10-15-6-2-1-3-7-15/h1-9,17-18,23H,10-14H2,(H,25,26)(H,28,29)/t17-,18-/m0/s1
SMILES OC(=O)CN1C(=O)[C@H](CCc2ccccc12)N[C@@H](CCc3ccccc3)C(=O)O
IUPAC Name (2S)-2-[[(3S)-1-(carboxymethyl)-2-oxo-4,5-dihydro-3H-1-benzazepin-3-yl]amino]-4-phenylbutanoic acid
Molecular Weight (g/mol) 396.44
Quantity 25 mg
Formula Weight 396.1685 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.