missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid), Mikromol™

Product Code. 17979584
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17979584 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17979584 Supplier Mikromol Supplier No. MM0621.010025
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
CAS 4271-30-1
Analyte or Component Names (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid (N-(4-Aminobenzoyl)-L-glutamic Acid)
Shipping Condition Room Temperature
Molecular Formula C12 H14 N2 O5
Certifications/Compliance ISO 17025
Synonym (2S)-2-[(4-Aminobenzoyl)amino]pentanedioic Acid, Folic Acid Imp. A (EP), N-(4-Aminobenzoyl)-L-glutamic Acid, Glutamic acid, N-(p-aminobenzoyl)- (6CI,7CI), Glutamic acid, N-(p-aminobenzoyl)-, L- (8CI), (4-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)-L-glutamic acid, (p-Aminobenzoyl)glutamic acid, N-(p-Aminobenzoyl)-L(+)-glutamic acid, N-(p-Aminobenzoyl)-L-glutamic acid, N-(p-Aminobenzoyl)glutamic acid, NSC 71042, Calcium Folinate Impurity A, Calcium Levofolinate Impurity A, Folic Acid Hydrate Impurity A, Methotrexate Impurity K, Folic Acid Impurity A
InChI Formula InChI=1S/C12H14N2O5/c13-8-3-1-7(2-4-8)11(17)14-9(12(18)19)5-6-10(15)16/h1-4,9H,5-6,13H2,(H,14,17)(H,15,16)(H,18,19)/t9-/m0/s1
SMILES Nc1ccc(cc1)C(=O)N[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-[(4-aminobenzoyl)amino]pentanedioic acid
Molecular Weight (g/mol) 266.25
Quantity 25 mg
Formula Weight 266.0903 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.