missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-Amino-4-[(RS)-methylsulfinyl]butanoic Acid (L-Methionine Sulfoxide), Mikromol™

Product Code. 17948644
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17948644 100 mg 100mg
17927584 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17948644 Supplier Mikromol Supplier No. MM0358.05
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-Amino-4-[(RS)-methylsulfinyl]butanoic Acid (L-Methionine Sulfoxide)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Methionine Sulfoxide
CAS 3226-65-1
Analyte or Component Names L-Methionine Sulfoxide
Shipping Condition Room Temperature
Molecular Formula C5 H11 N O3 S
Certifications/Compliance ISO 17025
Synonym L-Methionine S-oxide, Butanoic acid, 2-amino-4-(methylsulfinyl)-, (2S)- (9CI), Butyric acid, 2-amino-4-(methylsulfinyl)-, L- (8CI), 3: PN: WO2011158590 PAGE: 34 claimed protein, L-Methionine sulfoxide, Methionine sulfoxide, Methionine Impurity A
InChI Formula InChI=1S/C5H11NO3S/c1-10(9)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-,10?/m0/s1
SMILES CS(=O)CC[C@H](N)C(=O)O
IUPAC Name (2S)-2-amino-4-methylsulfinylbutanoic acid
Molecular Weight (g/mol) 165.21
Quantity 100 mg
Formula Weight 165.046 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.