missing translation for 'onlineSavingsMsg'
Learn More

(2S)-2-Hydroxy-2-phenylacetic Acid (L-Mandelic Acid), Mikromol™

Product Code. 17991456
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17991456 Supplier Mikromol Supplier No. MM1304.050025
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S)-2-Hydroxy-2-phenylacetic Acid (L-Mandelic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-(+)-Mandelic Acid
CAS 17199-29-0
Analyte or Component Names L-(+)-Mandelic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Certifications/Compliance ISO 17025
Synonym Benzeneacetic acid, α-hydroxy-, (αS)-, Benzeneacetic acid, α-hydroxy-, (S)-, Mandelic acid, L-(+)- (8CI), (αS)-α-Hydroxybenzeneacetic acid, (+)-(S)-Mandelic acid, (+)-Mandelic acid, (+)-α-Hydroxyphenylacetic acid, (2S)-2-Hydroxy-2-(phenyl)ethanoic acid, (S)-(+)-Hydroxyphenylacetic acid, (S)-(+)-Mandelic acid, (S)-2-Hydroxy-2-(phenyl)ethanoic acid, (S)-2-Hydroxy-2-phenylacetic acid, (S)-Mandelic acid, (S)-α-Hydroxybenzeneacetic acid, (S)-α-Hydroxyphenylacetic acid, L-(+)-Mandelic acid, L-Mandelic acid, Atomoxetine Hydrochloride Imp E (EP), Atomoxetine Hydrochloride Impurity E, Atomoxetine Impurity E
InChI Formula InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)/t7-/m0/s1
SMILES O[C@H](C(=O)O)c1ccccc1
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
Molecular Weight (g/mol) 152.15
Quantity 25 mg
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.