missing translation for 'onlineSavingsMsg'
Learn More

(2S,3S)-2-(4-Methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate Hydrochloride (N-Desmethyl-diltiazem Hydrochloride), Mikromol™

Product Code. 30460856
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460856 4 x 25 mg -
30052311 25 mg 25mg
17932236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460856 Supplier Mikromol Supplier No. MM0068.0300254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2S,3S)-2-(4-Methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate Hydrochloride (N-Desmethyl-diltiazem Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Desmethyldiltiazem Hydrochloride
CAS 130606-60-9
Analyte or Component Names N-Desmethyldiltiazem Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C21 H24 N2 O4 S . Cl H
Certifications/Compliance ISO 9001
Synonym Diltiazem Hydrochloride Imp. D (EP), Diltiazem Imp. D (EP), (2S,3S)-2-(4-Methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3,4,5-tetrahydro-1,5-benzothiazepin-3-yl Acetate Hydrochloride, Diltiazem Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethyl-diltiazem Hydrochloride, Diltiazem Hydrochloride Impurity D as Hydrochloride, Diltiazem Impurity D as Hydrochloride
InChI Formula InChI=1S/C21H24N2O4S.ClH/c1-14(24)27-19-20(15-8-10-16(26-3)11-9-15)28-18-7-5-4-6-17(18)23(21(19)25)13-12-22-2;/h4-11,19-20,22H,12-13H2,1-3H3;1H/t19-,20+;/m1./s1
Solution Type Neat
SMILES Cl.CNCCN1C(=O)[C@H](OC(=O)C)[C@@H](Sc2ccccc12)c3ccc(OC)cc3
IUPAC Name [(2S,3S)-2-(4-methoxyphenyl)-5-[2-(methylamino)ethyl]-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] acetate;hydrochloride
Molecular Weight (g/mol) 436.95
Quantity 4 x 25 mg
Formula Weight 436.1224 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.