missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydro-4H-1-benzopyran-4-one (Hesperetin), Mikromol™
Description
(2S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-2,3-dihydro-4H-1-benzopyran-4-one (Hesperetin), Mikromol
Specifications
Specifications
| Chemical Name or Material | Hesperetin |
| CAS | 520-33-2 |
| Analyte or Component Names | Hesperetin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 O6 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 4H-1-Benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, (2S)-, 4H-1-Benzopyran-4-one, 2,3-dihydro-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-, (S)-, Flavanone, 3',5,7-trihydroxy-4'-methoxy- (8CI), Hesperetin (6CI), (-)-Hesperetin, (2S)-2,3-Dihydro-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, 2,3-Dihydro-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one, 3',5,7-Trihydroxy-4'-methoxyflavanone, 5,7,3'-Trihydroxy-4'-methoxyflavanone, Eriodictyol 4'-monomethyl ether, Hesperitin, NSC 57654, Diosmin Imp. G (EP), Diosmin Impurity G |
| InChI Formula | InChI=1S/C16H14O6/c1-21-13-3-2-8(4-10(13)18)14-7-12(20)16-11(19)5-9(17)6-15(16)22-14/h2-6,14,17-19H,7H2,1H3/t14-/m0/s1 |
| Solution Type | Neat |
| SMILES | COc1ccc(cc1O)[C@@H]2CC(=O)c3c(O)cc(O)cc3O2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?