missing translation for 'onlineSavingsMsg'
Learn More

2Z-Dehydro Rosuvastatin sodium salt, TRC

Product Code. 30426533
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.50mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30426533 25 mg 25mg
30422447 2.5 mg 2.50mg
30426191 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426533 Supplier Toronto Research Chemicals Supplier No. TRCR70040025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2Z-Dehydro Rosuvastatin sodium salt
Analyte or Component Names 2Z-Dehydro Rosuvastatin sodium salt
Shipping Condition Room Temperature
Molecular Formula C22H25FN3NaO5S
Synonym (2Z,5S,6E)-7-[4-(4-fluorophenyl)-6-(1-methylethyl)-2-[methyl(methylsulfonyl)amino]-5-pyrimidinyl]-5-hydroxy-2,6-Heptadienoic acid
InChI Formula InChI=1S/C22H26FN3O5S.Na/c1-14(2)20-18(13-12-17(27)6-5-7-19(28)29)21(15-8-10-16(23)11-9-15)25-22(24-20)26(3)32(4,30)31;/h5,7-14,17,27H,6H2,1-4H3,(H,28,29);/q;+1/p-1/b7-5-,13-12+;/t17-;/m0./s1
Solution Type Neat
SMILES O[C@@H](C/C=C\C(O[Na])=O)/C=C/C(C(C1=CC=C(F)C=C1)=NC(N(C)[S](=O)(C)=O)=N2)=C2C(C)C
IUPAC Name sodium (S,2Z,6E)-7-(4-(4-fluorophenyl)-6-isopropyl-2-(N-methylmethylsulfonamido)pyrimidin-5-yl)-5-hydroxyhepta-2,6-dienoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 485.51
Quantity 25 mg
Formula Weight 485.14
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.