missing translation for 'onlineSavingsMsg'
Learn More

3-(1-Methylethyl)phenol, Mikromol™

Product Code. 17900706
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17900706

Brand: Mikromol MM0461.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-(1-Methylethyl)phenol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Isopropylphenol
CAS 618-45-1
Analyte or Component Names 3-Isopropylphenol
Shipping Condition Ice pack (-20°C)
Molecular Formula C9 H12 O
Certifications/Compliance ISO 17025
Synonym 3-(1-Methylethyl)phenol, Propofol Imp. F (EP), Propofol Impurity F
InChI Formula InChI=1S/C9H12O/c1-7(2)8-4-3-5-9(10)6-8/h3-7,10H,1-2H3
SMILES CC(C)c1cccc(O)c1
IUPAC Name 3-propan-2-ylphenol
Molecular Weight (g/mol) 136.19
Quantity 100 mg
Formula Weight 136.0888 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.