missing translation for 'onlineSavingsMsg'
Learn More

3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate, Mikromol™

Product Code. 18083300
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18083300 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18083300 Supplier Mikromol Supplier No. MM1497.010100
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[(1RS)-1-(Dimethylamino)ethyl]phenyl Dimethylcarbamate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R,S)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate
CAS 25081-93-0
Analyte or Component Names (R,S)-3-[1-(Dimethylamino)ethyl]phenyl dimethylcarbamate
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O2
Certifications/Compliance ISO 9001
Synonym N,N-Dimethylcarbamic acid 3-[1-(Dimethylamino)ethyl]phenyl ester, Dimethylcarbamic acid 3-[1-(Dimethylamino)ethyl]phenyl ester, Dimethylcarbamic acid m-[1-(Dimethylamino)ethyl]phenyl ester, m-[1-(Dimethylamino)ethyl]phenol dimethylcarbamate, Rivastigmine Related Compound B, Rivastigmine Hydrogen Tartrate Impurity B as Racemate, Rivastigmine Impurity B as Racemate
InChI Formula InChI=1S/C13H20N2O2/c1-10(14(2)3)11-7-6-8-12(9-11)17-13(16)15(4)5/h6-10H,1-5H3
SMILES CC(N(C)C)c1cccc(OC(=O)N(C)C)c1
IUPAC Name [3-[1-(dimethylamino)ethyl]phenyl] N,N-dimethylcarbamate
Molecular Weight (g/mol) 236.31
Quantity 100 mg
Formula Weight 236.1525 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.