missing translation for 'onlineSavingsMsg'
Learn More

3-(2-chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 11366935
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11366935 25 g 25g
10455712 1 g 1g
10273082 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11366935 Supplier Thermo Scientific Maybridge Supplier No. SB01884EB
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3919-74-2
Molecular Formula C11H7ClFNO3
Molecular Weight (g/mol) 255.63
MDL Number MFCD00055659
InChI Key REIPZLFZMOQHJT-UHFFFAOYSA-N
Synonym 3-2-chloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-1,2-oxazole-4-carboxylic acid, fcimic acid, 4-isoxazolecarboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxylic acid, 3-2-dihloro-6-fluorophenyl-5-methylisoxazole-4-carboxylic acid, 3-2-chloro-6-fluorophenyl-5-methyl-4-isoxazolecarboxic acid, pubchem8772, acmc-1ckjh, 3-2-chloro-6-fluorophenyl 5-methylisoxazole-4-carboxylic acid
PubChem CID 77520
IUPAC Name 3-(2-chloro-6-fluorophenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=C(C(O)=O)C(=NO1)C1=C(F)C=CC=C1Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 255.63
Percent Purity 97%
Chemical Name or Material 3-(2-Chloro-6-fluorophenyl)-5-methylisoxazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.