missing translation for 'onlineSavingsMsg'
Learn More

3-[(2S)-Piperidin-2-yl]pyridine (Anabasine), Mikromol™

Product Code. 18043880
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18043880 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18043880 Supplier Mikromol Supplier No. MM0517.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-[(2S)-Piperidin-2-yl]pyridine (Anabasine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(2S)-Piperidin-2-yl]pyridine (Anabasine)
CAS 494-52-0
Analyte or Component Names 3-[(2S)-Piperidin-2-yl]pyridine (Anabasine)
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2
Certifications/Compliance ISO 9001
Synonym Nicotine Imp. G (EP), Nicotine USP Related Compound G, Nicotine USP RC G, 3-[(2S)-Piperidin-2-yl]pyridine, Anabasine, Nicotine Related Compound G, Nicotine Impurity G
InChI Formula InChI=1S/C10H14N2/c1-2-7-12-10(5-1)9-4-3-6-11-8-9/h3-4,6,8,10,12H,1-2,5,7H2/t10-/m0/s1
SMILES C1CC[C@H](NC1)c2cccnc2
IUPAC Name 3-[(2S)-piperidin-2-yl]pyridine
Molecular Weight (g/mol) 162.23
Quantity 100 mg
Formula Weight 162.1157 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.