missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol, LoGiCal

Product Code. 30457864
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457864 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457864 Supplier LoGiCal Supplier No. LGCFOR0027.06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol
CAS 62572-90-1
Analyte or Component Names 3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol
Shipping Condition Room Temperature
Molecular Formula C12 H18 O4
Certifications/Compliance ISO 9001
Synonym Metoprolol Succinate Imp. D (EP), Metoprolol Tartrate Imp. D (EP), Metoprolol Imp. D (EP), (2RS)-3-[4-(2-Methoxyethyl)phenoxy]propane-1,2-diol, Metoprolol Succinate Impurity D, Metoprolol Tartrate Impurity D, Metoprolol Impurity D, Metoprolol Impurity D
InChI Formula InChI=1S/C12H18O4/c1-15-7-6-10-2-4-12(5-3-10)16-9-11(14)8-13/h2-5,11,13-14H,6-9H2,1H3
Solution Type Neat
SMILES COCCc1ccc(OCC(O)CO)cc1
IUPAC Name 3-[4-(2-methoxyethyl)phenoxy]propane-1,2-diol
Industry Type Clinical Testing
Molecular Weight (g/mol) 226.27
Quantity 10 mg
Formula Weight 226.1205 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.