missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzylamine hydrobromide, 98%, Thermo Scientific™

Product Code. 11424877
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424877 1 g 1g
11434877 5 g 5g
11444877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11424877 Supplier Thermo Scientific Alfa Aesar Supplier No. B21833.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16290-26-9
Molecular Formula C7H10BrNO2
Molecular Weight (g/mol) 220.07
MDL Number MFCD00012859
InChI Key BVFZTXFCZAXSHN-UHFFFAOYSA-N
Synonym 3,4-dihydroxybenzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol hydrobromide, dhba hydrobromide, acmc-1afa4, 1, 4-aminomethyl-, hydrobromide, 4-aminomethyl catechol hydrobromide, 3,4 dihydroxybenzylamine hydrobromide, 3,4-dihy-droxybenzylamine hydrobromide, 3,4-dihydroxy benzylamine hydrobromide, 4-aminomethyl benzene-1,2-diol, bromide
PubChem CID 13343562
SMILES [H+].[Br-].NCC1=CC=C(O)C(O)=C1

Specifications

Melting Point 196°C to 200°C
Quantity 1 g
Beilstein 4002646
Sensitivity Hygroscopic
IUPAC Name 4-(aminomethyl)benzene-1,2-diol;hydrobromide
Formula Weight 220.07
Percent Purity 98%
Chemical Name or Material 3,4-Dihydroxybenzylamine hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.