missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluoro-3-methoxyphenyl)propionic acid, 96%, Thermo Scientific™

Product Code. 15418695
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418695 250 mg 250mg
15486777 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418695 Supplier Thermo Scientific Alfa Aesar Supplier No. H33894.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864960-96-3
Molecular Formula C10H11FO3
Molecular Weight (g/mol) 198.19
MDL Number MFCD11655589
InChI Key ANPCCWWNYXSFDQ-UHFFFAOYSA-N
Synonym 3-4-fluoro-3-methoxyphenyl propanoic acid, 3-4-fluoro-3-methoxyphenyl propionic acid, benzenepropanoic acid, 4-fluoro-3-methoxy, 3-4-fluoro-3-methoxy-phenyl-propionic acid, 3-4-fluoro-3-methoxyphenyl propanoic acid im ocs
PubChem CID 21098606
IUPAC Name 3-(4-fluoro-3-methoxyphenyl)propanoic acid
SMILES COC1=C(F)C=CC(CCC(O)=O)=C1

Specifications

Melting Point 90°C to 94°C
Quantity 250 mg
Formula Weight 198.19
Percent Purity 96%
Chemical Name or Material 3-(4-Fluoro-3-methoxyphenyl)propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.