missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Hydroxyphenyl)propionic acid, 98%

Product Code. 10467310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10467310 25 g Glass bottle 25g
10100272 5 g Glass bottle 5g
10404945 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10467310 Supplier Thermo Scientific Acros Supplier No. 121750250
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501-97-3 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-97-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002778
InChI Key NMHMNPHRMNGLLB-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propionic acid, phloretic acid, 3-4-hydroxyphenyl propanoic acid, desaminotyrosine, hydro-p-coumaric acid, dihydro-p-coumaric acid, benzenepropanoic acid, 4-hydroxy, 4-hydroxyphenylpropionic acid, p-hydroxyhydrocinnamic acid, 3-p-hydroxyphenyl propionic acid
PubChem CID 10394
ChEBI CHEBI:32980
IUPAC Name 3-(4-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC=C1CCC(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126°C to 130°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2CO2H
Quantity 25 g
Beilstein 10, 244
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in alchol,acetone and ether
Formula Weight 166.18
Percent Purity 98%
Physical Form Powder or Granules
Chemical Name or Material 3-(4-Hydroxyphenyl)propionic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-(4-Hydroxyphenyl)propionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.