missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(tert-Butyl)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole, Tech., Thermo Scientific™

Product Code. 10180882
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10180882 1 g Amber Glass Bottle 1g
10364722 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10180882 Supplier Thermo Scientific Maybridge Supplier No. RF00728DA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 175204-40-7
Molecular Formula C13H15ClN2O
Molecular Weight (g/mol) 250.726
MDL Number MFCD00124908
InChI Key ZLQQUIHBQATFOP-UHFFFAOYSA-N
Synonym 3-4-tert-butylphenyl-5-chloromethyl-1,2,4-oxadiazole, 3-4-tert-butyl phenyl-5-chloromethyl-1,2,4-oxadiazole, 3-4-tert-butylphenyl-5-chloromethyl-1,2,4 oxadiazole, 1,2,4-oxadiazole,5-chloromethyl-3-4-1,1-dimethylethyl phenyl, 3-4-tert-butyl-phenyl-5-chloromethyl-1,2,4 o xadiazole
PubChem CID 2781701
IUPAC Name 3-(4-tert-butylphenyl)-5-(chloromethyl)-1,2,4-oxadiazole
SMILES CC(C)(C)C1=CC=C(C=C1)C2=NOC(=N2)CCl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 250.73
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-[4-(Tert-butyl)phenyl]-5-(chloromethyl)-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.