missing translation for 'onlineSavingsMsg'
Learn More

3,5-Bis(trifluoromethyl)benzyl chloride, 97%, Thermo Scientific™

Product Code. 11447877
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11447877 5 g 5g
11437877 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447877 Supplier Thermo Scientific Alfa Aesar Supplier No. A16553.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3,5-Bis(trifluoromethyl)benzyl chloride has been used in the preparation of 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene. 1-(azidomethyl)-3,5-bis-(trifluoromethyl)benzene, in 94% yield by an efficient nucleophilic substitution reaction between 3,5-bis-(trifluoromethyl)benzyl chloride, 4, and sodium azide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75462-59-8
Molecular Formula C9H5ClF6
Molecular Weight (g/mol) 262.579
MDL Number MFCD00009906
InChI Key OINTXXMBRBLMHH-UHFFFAOYSA-N
Synonym 3,5-bis trifluoromethyl benzyl chloride, 1-chloromethyl-3,5-bis trifluoromethyl benzene, 3,5-bis-trifluoromethyl benzyl chloride, 3,5-bis trifluoromethyl benzylchloride, 3,5-di trifluoromethyl benzyl chloride, benzene, 1-chloromethyl-3,5-bis trifluoromethyl, 3,5-bis trifluoromethyl-1-chloromethyl benzene, 1,3-bis trifluoromethyl-5-chloromethyl benzene
PubChem CID 123576
IUPAC Name 1-(chloromethyl)-3,5-bis(trifluoromethyl)benzene
SMILES C1=C(C=C(C=C1C(F)(F)F)C(F)(F)F)CCl

Specifications

Melting Point 29°C to 33°C
Boiling Point 46°C (3 mmHg)
Flash Point 71°C (161°F)
Quantity 5 g
UN Number UN3261
Beilstein 656502
Solubility Information Slightly soluble in water.
Formula Weight 262.58
Percent Purity 97%
Chemical Name or Material 3,5-Bis(trifluoromethyl)benzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.