missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloro-4-hydroxybenzoic acid, 97%

Product Code. 11489743
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489743 100 g 100g
11469743 5 g 5g
11479743 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489743 Supplier Thermo Scientific Alfa Aesar Supplier No. A12923.22
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3336-41-2 | C7H4Cl2O3 | 207.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3336-41-2
Molecular Formula C7H4Cl2O3
Molecular Weight (g/mol) 207.01
MDL Number MFCD00002550
InChI Key AULKDLUOQCUNOK-UHFFFAOYSA-N
Synonym diclhbz, benzoic acid, 3,5-dichloro-4-hydroxy, 3,5-dichloro-4-hydroxy-benzoic acid, pubchem3689, acmc-1cjos, epitope id:122960, 3,5-dichloro-p-salicylic acid, rarechem al be 0933, timtec-bb sbb042792, benzoic acid,5-dichloro-4-hydroxy
PubChem CID 18749
ChEBI CHEBI:53685
IUPAC Name 3,5-dichloro-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC(Cl)=C(O)C(Cl)=C1

Specifications

Melting Point 265°C to 270°C
Quantity 100 g
Beilstein 2616297
Formula Weight 207.01
Percent Purity 97%
Chemical Name or Material 3,5-Dichloro-4-hydroxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.