missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluoroanisole, 98%, Thermo Scientific™

Product Code. 10179272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10179272 1 g Glass Bottle 1g
10712091 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10179272 Supplier Thermo Scientific Acros Supplier No. 249560010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93343-10-3
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00042560
InChI Key OTGQPYSISUUHAF-UHFFFAOYSA-N
Synonym 3,5-difluoroanisole, 3,5-difluoro-1-methoxybenzene, benzene, 1,3-difluoro-5-methoxy, 1,3-difluoro-5-methoxy-benzene, 3,5-difluoro anisole, 3,5-difluoroanisol, 3.5-difluoroanisole, pubchem1967, 3,5-difluoroanisole;, 3,5,-difluoroanisole;
PubChem CID 2724518
IUPAC Name 1,3-difluoro-5-methoxybenzene
SMILES COC1=CC(F)=CC(F)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.23g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula F2C6H3OCH3
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Quantity 1 g
Formula Weight 144.12
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 3, 5-Difluoroanisole, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,5-Difluoroanisole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.