missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxybenzonitrile, 97%

Product Code. 15486727
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15486727 1 g 1g
15406737 250 mg 250mg
15496727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15486727 Supplier Thermo Scientific Alfa Aesar Supplier No. H33856.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14543-43-2 | C7H6N2O | 134.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14543-43-2
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.138
MDL Number MFCD00234273
InChI Key ZEWCASRNRWXXSO-UHFFFAOYSA-N
PubChem CID 271258
IUPAC Name 3-amino-4-hydroxybenzonitrile
SMILES C1=CC(=C(C=C1C#N)N)O

Specifications

Melting Point 151°C to 153°C
Quantity 1 g
Sensitivity Air Sensitive
Synonym 2-amino-4-cyanophenol, 2-amino-4-cyano-phenol, benzonitrile, 3-amino-4-hydroxy, 3-amino-4-hydroxybenzenecarbonitrile, 5-cyano-2-hydroxyaniline, acmc-209cv1, 3-amino-4-hydroxy-benzonitrile, benzonitrile,3-amino-4-hydroxy, 5-cyano-2-hydroxy-aniline
Formula Weight 134.14
Percent Purity 97%
Chemical Name or Material 3-Amino-4-hydroxybenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.