missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzoic Acid, Mikromol™

Product Code. 30051813
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051813 100 mg 100mg
30049071 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051813 Supplier Mikromol Supplier No. MM0217.07
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Aminobenzoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Aminobenzoic acid
CAS 99-05-8
Analyte or Component Names 3-Aminobenzoic acid
Shipping Condition Room Temperature
Molecular Formula C7 H7 N O2
Certifications/Compliance ISO 17025
Synonym 3-Aminobenzoic Acid, Mesalazine Imp. D (EP), Mesalazine Impurity D
InChI Formula InChI=1S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H,9,10)
SMILES Nc1cccc(c1)C(=O)O
IUPAC Name 3-aminobenzoic acid
Molecular Weight (g/mol) 137.14
Quantity 100 mg
Formula Weight 137.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.