missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-hydroxybenzoic acid, 97%

Product Code. p-7025701
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11489257

Brand: Thermo Scientific Alfa Aesar B21338.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14348-41-5 | C7H5BrO3 | 217.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14348-41-5
Molecular Formula C7H5BrO3
Molecular Weight (g/mol) 217.02
MDL Number MFCD00017547
InChI Key XMEQDAIDOBVHEK-UHFFFAOYSA-N
Synonym 3-bromo-4-hydroxy-benzoic acid, benzoic acid, 3-bromo-4-hydroxy, acmc-20aix6, 3-bromo-4-hydroxybenzoicacid, 3-bromo-4-hydroxybenzoic acid, 3-bromo-4-hydroxybenzoicacidhydrate, 3-bromo-4-hydroxybenzoic acid 10g
PubChem CID 84368
IUPAC Name 3-bromo-4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C(Br)=C1

Specifications

Melting Point 176°C to 178°C
Quantity 10 g
Formula Weight 217.02
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-hydroxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.