missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)phenoxyacetic acid, 97%

Product Code. 11354885
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354885 1 g 1g
11364885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354885 Supplier Thermo Scientific Alfa Aesar Supplier No. L18029.03
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Læg i indkøbskurv
Request Bulk or Custom Format
Denne vare kan ikke returneres. Se returpolicy

CAS: 136645-25-5 | C9H9BrO3 | 245.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 136645-25-5
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD02093985
InChI Key MSTODKGDFXWAIO-UHFFFAOYSA-N
Synonym 3-bromomethyl phenoxyacetic acid, 2-3-bromomethyl phenoxy acetic acid, acmc-1c8fy
PubChem CID 7010327
IUPAC Name 2-[3-(bromomethyl)phenoxy]acetic acid
SMILES C1=CC(=CC(=C1)OCC(=O)O)CBr

Tekniske data

Melting Point 124°C to 128°C
Quantity 1 g
UN Number UN3261
Formula Weight 245.07
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)phenoxyacetic acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.