missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-iodoaniline, 98%, Thermo Scientific™

Product Code. 11402397
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11402397 25 g 25g
11492387 5 g 5g
11412397 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402397 Supplier Thermo Scientific Alfa Aesar Supplier No. A14256.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-4-iodoaniline used as a intermediate for organic syntheses.

Notes
Light sensitive. Store in dark. Store in cool, dry place in tightly closed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135050-44-1
Molecular Formula C6H5ClIN
Molecular Weight (g/mol) 253.47
MDL Number MFCD00051845
InChI Key ONZHMGRKWJMTDE-UHFFFAOYSA-N
Synonym benzenamine, 3-chloro-4-iodo, 3-chloro-4-iodobenzenamine, 4-amino-2-chloroiodobenzene, 3-chloro-4-iodophenylamine, pubchem3812, 3-chloro4-iodoaniline, zlchem 1106, acmc-209bxp, 3-chloro-4-iodo-aniline, 3-chloro-4-iodo aniline
PubChem CID 282926
IUPAC Name 3-chloro-4-iodoaniline
SMILES NC1=CC=C(I)C(Cl)=C1

Specifications

Melting Point 65°C to 71°C
Quantity 25 g
UN Number UN2811
Beilstein 3236464
Sensitivity Light sensitive
Formula Weight 253.47
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-iodoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.