missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoic acid, 99%

Product Code. 11454223
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454223 25 g 25g
11464223 100 g 100g
11474223 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454223 Supplier Thermo Scientific Alfa Aesar Supplier No. A10876.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 535-80-8 | C7H5ClO2 | 156.565 g/mol

A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A Pseudomonas species which utilized 3-chlorobenzoic acid as a carbon source converted this compound to 3-hydroxybenzoic and 2, 5-dihydroxybenzoic acids. A methanogenic consortium able to use 3-chlorobenzoic acid as its sole energy and carbon source was enriched from anaerobic sewage sludge. A Pseudomonas putida strain 87 capable of assimilating 3-chlorobenzoic acid as a sole source of carbon and energy (3Cba+) was isolated. coupling of 3-chlorobenzoic acid with phenylboronic acid proceeds at room temperature using 0.5 % Pd and at 100°C using 0.1 % Pd to provide the coupled product in 97 % yeild.

Solubility
Slioghtly soluble in water(0.45 g/L ).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-80-8
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD00002491
InChI Key LULAYUGMBFYYEX-UHFFFAOYSA-N
Synonym m-chlorobenzoic acid, benzoic acid, 3-chloro, benzoic acid, m-chloro, 3-chlorobenzoicacid, 3-chloro-benzoic acid, m-chlorobenzoate, acido m-clorobenzoico, unii-02uoj7064k, mcba, ccris 5992
PubChem CID 447
ChEBI CHEBI:49410
IUPAC Name 3-chlorobenzoic acid
SMILES C1=CC(=CC(=C1)Cl)C(=O)O

Specifications

Melting Point 154°C to 160°C
Boiling Point 274°C to 276°C
Odor Odorless
Quantity 25 g
Beilstein 907218
Merck Index 14,2123
Solubility Information Slioghtly soluble in water(0.45g/L ).
Formula Weight 156.57
Percent Purity 99%
Chemical Name or Material 3-Chlorobenzoic acid
compliance-icons
Product Identifier
  • 3-Chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Chlorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.