missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. p-7040940
Click to view available options
Quantity:
0.5 g
10 g
Unit Size:
0.5g
10g
This item is not returnable. View return policy

Product Code. 15427358

Brand: Thermo Scientific Alfa Aesar H60854.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Chloromethyl)benzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31719-77-4
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00191922
InChI Key PBSUMBYSVFTMNG-UHFFFAOYSA-N
Synonym 3-chloromethyl benzoic acid, alpha-chloro-m-toluic acid, 3-chloromethyl-benzoic acid, benzoic acid, 3-chloromethyl, pubchem7446, acmc-209hog, 3-carboxybenzyl chloride, 3-chloromethyl benzoicacid, ksc497k4l, chembl67150
PubChem CID 241752
IUPAC Name 3-(chloromethyl)benzoic acid
SMILES OC(=O)C1=CC=CC(CCl)=C1

Specifications

Melting Point 136°C to 138°C
Quantity 0.5 g
UN Number UN3261
Solubility Information Soluble in methanol.
Formula Weight 170.59
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)benzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.