missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GOLPH3 Polyclonal specifically detects GOLPH3 in Human∣Mouse samples. It is validated for Immunoprecipitation∣Western Blot.
Specifications
Specifications
| Antigen | GOLPH3 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 405 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Coat protein GPP34, FLJ90675, Golgi phosphoprotein 3, golgi phosphoprotein 3 (coat-protein), golgi protein, golgi-associated protein, GOPP1, GPP34coat-protein, MIDASgolgi peripheral membrane protein 1, 34 kDa, Mitochondrial DNA absence factor |
| Host Species | Rabbit |
| Immunogen | Recognizes region between residues Between between 1 and 50.. |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?