missing translation for 'onlineSavingsMsg'
Learn More

3’’’-Epi-Empagliflozin, TRC

Product Code. 30011511
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011511 25 mg 25mg
30007068 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011511 Supplier Toronto Research Chemicals Supplier No. TRCE58600025MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864070-43-9
Molecular Formula C23H27ClO7
Molecular Weight (g/mol) 450.91
Synonym (1S)-1,5-Anhydro-1-C-[4-chloro-3-[[4-[[(3R)-tetrahydro-3-furanyl]oxy]phenyl]methyl]phenyl]-D-glucitol, Empagliflozin Impurity C
IUPAC Name (2S,3R,4R,5S,6R)-2-(4-chloro-3-(4-(((R)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
SMILES OC[C@H]1O[C@@H](C2=CC(CC3=CC=C(O[C@H]4COCC4)C=C3)=C(Cl)C=C2)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Analyte or Component Names 3' ' '-Epi-Empagliflozin
Shipping Condition Room Temperature
Solution Type Neat
Industry Type Forensics
Quantity 25 mg
Percent Purity >95%
Chemical Name or Material 3' ' '-Epi-Empagliflozin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.