missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-5-methylbenzyl bromide, 97%, Thermo Scientific™

Product Code. 11372649
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372649 1 g 1g
11382649 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372649 Supplier Thermo Scientific Alfa Aesar Supplier No. H26702.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Fluoro-5-methylbenzyl bromide is used for the production of inorganic bromides. And also used in pharmaceuticals intermediates.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212268-39-8
Molecular Formula C8H8BrF
Molecular Weight (g/mol) 203.05
MDL Number MFCD03094316
InChI Key PVDKNKUVHSULSN-UHFFFAOYSA-N
Synonym 3-fluoro-5-methylbenzyl bromide, 1-bromomethyl-3-fluoro-5-methylbenzene, 3-?fluoro-?5-?methylbenzyl bromide, 3-bromomethyl-1-fluoro-5-methylbenzene, 5-bromomethyl-1-fluoro-3-methylbenzene
PubChem CID 2778451
IUPAC Name 1-(bromomethyl)-3-fluoro-5-methylbenzene
SMILES CC1=CC(CBr)=CC(F)=C1

Specifications

Quantity 1 g
UN Number UN3265
Solubility Information Difficult to mix.
Formula Weight 203.06
Percent Purity 97%
Chemical Name or Material 3-Fluoro-5-methylbenzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.