missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorocatechol, 99%

Product Code. p-7026240
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11449667

Brand: Thermo Scientific Alfa Aesar B22145.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 363-52-0 | C6H5FO2 | 128.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 363-52-0
Molecular Formula C6H5FO2
Molecular Weight (g/mol) 128.10
MDL Number MFCD00042582
InChI Key DXOSJQLIRGXWCF-UHFFFAOYSA-N
Synonym 3-fluorocatechol, pyrocatechol, 3-fluoro, 1,2-benzenediol, 3-fluoro, 1,2-dihydroxy-3-fluorobenzene, 3-fluoro-1,2-dihydroxybenzene, 3-fluoro-1,2-benzenediol, 1-fluoro-2,3-dihydroxybenzene, 3fa, pubchem4142
PubChem CID 67764
ChEBI CHEBI:39876
IUPAC Name 3-fluorobenzene-1,2-diol
SMILES OC1=CC=CC(F)=C1O

Specifications

Melting Point 71°C to 73°C
Quantity 1 g
UN Number UN1759
Beilstein 2436074
Formula Weight 128.1
Percent Purity 99%
Chemical Name or Material 3-Fluorocatechol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.