missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorotoluene, 98%

Product Code. 11389479
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389479 100 g 100g
11379479 25 g 25g
11463137 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11389479 Supplier Thermo Scientific Alfa Aesar Supplier No. A16107.22
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 352-70-5 | C7H7F | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 352-70-5
Molecular Formula C7H7F
Molecular Weight (g/mol) 110.13
MDL Number MFCD00000339
InChI Key BTQZKHUEUDPRST-UHFFFAOYSA-N
Synonym 3-fluorotoluene, m-fluorotoluene, benzene, 1-fluoro-3-methyl, 1-fluoro-3-methyl-benzene, 1-methyl-3-fluorobenzene, toluene, m-fluoro, unii-f72w445dq0, 3-fluorobenzyl radical, meta-fluorotoluene, acmc-209ifn
PubChem CID 9606
IUPAC Name 1-fluoro-3-methylbenzene
SMILES CC1=CC=CC(F)=C1

Specifications

Melting Point -87°C
Density 0.993
Boiling Point 114°C to 116°C
Flash Point 9°C (48°F)
Odor Aromatic
Refractive Index 1.469
Quantity 100 g
UN Number UN2388
Beilstein 1903631
Merck Index 14,4180
Formula Weight 110.13
Percent Purity 98%
Chemical Name or Material 3-Fluorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.