missing translation for 'onlineSavingsMsg'
Learn More

3-((Furan-2-ylmethyl)imino]-N,N-dimethyl-3H-1,2,4-dithiazol-5-amine Hydroiodide, TRC

Product Code. 30395801
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395801 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395801 Supplier Toronto Research Chemicals Supplier No. TRCF600695250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(Furan-2-ylmethyl)imino]-N,N-dimethyl-3H-1,2,4-dithiazol-5-amine Hydroiodide
Analyte or Component Names 3-[(Furan-2-ylmethyl)imino]-N,N-dimethyl-3H-1,2,4-dithiazol-5-amine Hydroiodide
Shipping Condition Room Temperature
Molecular Formula C9H11N3OS2 . HI
InChI Formula InChI=1S/C9H11N3OS2.HI/c1-12(2)9-11-8(14-15-9)10-6-7-4-3-5-13-7;/h3-5H,6H2,1-2H3;1H
Solution Type Neat
SMILES CN(C)C(SS/1)=NC1=N/CC2=CC=CO2.I
Industry Type Pharmaceutical
Molecular Weight (g/mol) 241.3312791
Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.