missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2-naphthoic acid, 98%

Product Code. 10152200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152200 5 g Glass bottle 5g
10725571 100 g Plastic bottle 100g
10337140 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10152200 Supplier Thermo Scientific Acros Supplier No. 121610050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 92-70-6 | C11H8O3 | 188.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-70-6
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.18
MDL Number MFCD00004103
InChI Key ALKYHXVLJMQRLQ-UHFFFAOYSA-N
Synonym 3-hydroxy-2-naphthoic acid, 2-hydroxy-3-naphthoic acid, bon acid, bona, developer bon, 2-naphthol-3-carboxylic acid, 2-naphthalenecarboxylic acid, 3-hydroxy, b.o.n. acid, c.i. developer 20, miketazol developer ons
PubChem CID 7104
ChEBI CHEBI:80383
IUPAC Name 3-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 217°C to 223°C
Color Beige-Brown to Yellow
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 10, 333
Merck Index 15, 4874
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alkalies,freely soluble in alcohol,ether and benzene
Formula Weight 188.18
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3-Hydroxy-2-naphthoic acid, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Hydroxy-2-naphthoic acid
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.