missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-4-methoxybenzoic acid, 99%

Product Code. 10148742 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10148742 5 g Glass bottle 5g
10552031 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10148742 Supplier Thermo Scientific Acros Supplier No. 205030050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 645-08-9 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645-08-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002507
InChI Key LBKFGYZQBSGRHY-UHFFFAOYSA-N
Synonym isovanillic acid, acide isovanillique, 3-hydroxyanisic acid, 3-hydroxy-p-anisic acid, benzoic acid, 3-hydroxy-4-methoxy, p-anisic acid, 3-hydroxy, chembl88700, 3-hydroxy-4-methoxy-benzoic acid, 3-hydroxy-4-methoxybenzoicacid, 5-carboxyguaiacol
PubChem CID 12575
ChEBI CHEBI:63798
IUPAC Name 3-hydroxy-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 245°C to 250°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CO2H
Quantity 5 g
Beilstein 10, 393
Formula Weight 168.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3-Hydroxy-4-methoxybenzoic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Hydroxy-4-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.