missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxybenzophenone, 98+%

Product Code. 11489206
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
missing translation for 'unitSize'
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489206 1 g 1g
11499206 5 g 5g
11409216 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11489206 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A18061.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13020-57-0 | C13H10O2 | 198.221 g/mol

3-Hydroxybenzophenone is a benzophenone metabolite with potential estrogenic and anti-androgenic activity. is a more attractive hydrogen bonding co-material for MCPBA, as the 3-hydroxyl group is unable to form an intramolecular hydrogen bond. It is commonly used as a radical and cationic photo initiators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxybenzophenone is a benzophenone metabolite with potential estrogenic and anti-androgenic activity. is a more attractive hydrogen bonding co-material for MCPBA, as the 3-hydroxyl group is unable to form an intramolecular hydrogen bond. It is commonly used as a radical and cationic photo initiators.

Solubility
Soluble in Chloroform, DCM. Ethyl Acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13020-57-0
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.221
MDL Number MFCD00002297
InChI Key SHULEACXTONYPS-UHFFFAOYSA-N
Synonym 3-hydroxybenzophenone, 3-hydroxyphenyl phenyl methanone, m-hydroxybenzophenone, methanone, 3-hydroxyphenyl phenyl, 3-hydroxy-benzophenon, dsstox_cid_27856, dsstox_rid_82609, dsstox_gsid_47880, 3-hydroxyphenyl phenyl ketone
PubChem CID 83050
IUPAC Name (3-hydroxyphenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)O

Specifications

Melting Point 113°C to 115°C
Quantity 1 g
Solubility Information Soluble in Chloroform,DCM. Ethyl Acetate.
Formula Weight 198.22
Percent Purity 99%
Chemical Name or Material 3-Hydroxybenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.