missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzophenone, Mikromol™

Product Code. 17922076
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17922076 Supplier Mikromol Supplier No. MM0001.13

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3-Methylbenzophenone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methylbenzophenone
CAS 643-65-2
Analyte or Component Names 3-Methylbenzophenone
Shipping Condition Room Temperature
Molecular Formula C14 H12 O
Certifications/Compliance ISO 9001
Synonym 3-Methylbenzophenone
InChI Formula InChI=1S/C14H12O/c1-11-6-5-9-13(10-11)14(15)12-7-3-2-4-8-12/h2-10H,1H3
SMILES Cc1cccc(c1)C(=O)c2ccccc2
IUPAC Name (3-methylphenyl)-phenylmethanone
Molecular Weight (g/mol) 196.24
Quantity 100 mg
Formula Weight 196.0888 g/mol
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.