missing translation for 'onlineSavingsMsg'
Learn More

3-[(tert-Butoxycarbonyl)amino]-5-hydroxybenzoic acid, 95%, Thermo Scientific™

Product Code. 10732774
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10732774 1 g Amber Glass Bottle 1g
11396895 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10732774 Supplier Thermo Scientific Maybridge Supplier No. MO08032DA
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 232595-59-4
Molecular Formula C12H15NO5
Molecular Weight (g/mol) 253.25
MDL Number MFCD02682202
InChI Key KSTCXQVSGVVIAO-UHFFFAOYSA-N
Synonym 3-tert-butoxycarbonyl amino-5-hydroxybenzoic acid, 3-n-boc-amino-5-hydroxybenzoic acid, 3-amino-5-hydroxybenzoic acid, n-boc protected, 3-tert-butoxycarbonylamino-5-hydroxy-benzoic acid, 3-tert-butoxy carbonylamino-5-hydroxybenzoic acid
PubChem CID 18322886
IUPAC Name 3-hydroxy-5-[(2-methylpropan-2-yl)oxycarbonylamino]benzoic acid
SMILES CC(C)(C)OC(=O)NC1=CC(O)=CC(=C1)C(O)=O

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.25
Percent Purity 95%
Chemical Name or Material 3-[(Tert-butoxycarbonyl)amino]-5-hydroxybenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.